missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AGL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565863
This item is not returnable.
View return policy
Description
Immunogen sequence: LAKCLGPFDEW ESRLRVAKES GYNMIHFTPL QTLGLSRSCY SLANQLELNP DFSRPNRKYT WNDVGQLVEK LKKEWNVICI TDVVYNHTAA NSKWIQEHPE Highest antigen sequence identity to the following orthologs - mouse 90%, rat 92%.
This gene encodes the glycogen debrancher enzyme which is involved in glycogen degradation. This enzyme has two independent catalytic activities which occur at different sites on the protein: a 4-alpha-glucotransferase activity and a amylo-1,6-glucosidase activity. Mutations in this gene are associated with glycogen storage disease although a wide range of enzymatic and clinical variability occurs which may be due to tissue-specific alternative splicing. Alternatively spliced transcripts encoding different isoforms have been described.
Specifications
| AGL | |
| Polyclonal | |
| Unconjugated | |
| AGL | |
| 1-Apr; 4-alpha-glucanotransferase; 4-glucantransferase; AGL; amylo-1, 6-glucosidase, 4-alpha-glucanotransferase; Amylo-1,6-glucosidase; amylo-alpha-1, 6-glucosidase, 4-alpha-glucanotransferase; amylo-alpha-1, 6-glucosidase, 4-alpha-glucanotransferaseprovided; Amylo-alpha-1,6-glucosidase; Dextrin 6-alpha-D-glucosidase; GDE; glycogen debrancher; Glycogen debranching enzyme; glycogen debranching protein; Oligo-1; Oligo-1,4-1,4-glucantransferase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 178 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P35573 | |
| AGL | |
| Recombinant Human AGL. Recombinant protein control fragment (Product #RP-106531). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction