missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AGMAT Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA555819

Product Code. 15703741

  • € 534.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: RRCVDEGLLD CKRVVQIGIR GSSTTLDPYR YNRSQGFRVV LAEDCWMKSL VPLMGEVRQQ MGGKPIYISF DI Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 88%.

AGMAT (Agmatinase) hydrolyzes agmatine to putrescine and urea. It is highly expressed in liver and kidney and is also found in skeletal muscle, fetal liver, brain, testis, skin and the gastrointestinal tract.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

AGMAT
Polyclonal
Unconjugated
AGMAT
5033405N08Rik; AGMAT; agmatinase; agmatinase, mitochondrial; Agmatine ureohydrolase; agmatine ureohydrolase (agmatinase); AUH
Rabbit
Antigen affinity chromatography
RUO
79814
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9BSE5
AGMAT
Recombinant protein corresponding to Human AGMAT. Recombinant protein control fragment (Product #RP-94090).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.