missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ago2/eIF2C2 Antibody (036), Janelia Fluor™ 646, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90675JF646
This item is not returnable.
View return policy
Description
Ago2/eIF2C2 Monoclonal specifically detects Ago2/eIF2C2 in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Ago2/eIF2C2 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 36 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| AGO2argonaute 2, argonaute2, EC 3.1.26.n1, EC 3.1.26.n2, eIF2C 2, eIF-2C 2, Eukaryotic translation initiation factor 2C 2, eukaryotic translation initiation factor 2C, 2, hAgo2, MGC3183, PAZ Piwi domain protein, PPD, protein argonaute-2, Protein slicer, Q10 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Ago2/eIF2C2 (Q8CJG0; Met1-Ala860). | |
| 0.1 mL | |
| Epigenetics | |
| 27161 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction