Learn More
Ago2/eIF2C2 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99647APC
This item is not returnable.
View return policy
Description
Ago2/eIF2C2 Polyclonal antibody specifically detects Ago2/eIF2C2 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| Immunocytochemistry | |
| Store at 4C in the dark. | |
| Purified |