missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AGPAT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117051
This item is not returnable.
View return policy
Description
Positive Samples: 293T, SW620, Mouse kidney, Rat kidney Immunogen sequence: PIVPIVMSSY QDFYCKKERR FTSGQCQVRV LPPVPTEGLT PDDVPALADR VRHSMLTVFR EISTDGRGGG DYLKKPGGGG.
This gene encodes an enzyme that converts lysophosphatidic acid (LPA) into phosphatidic acid (PA). LPA and PA are two phospholipids involved in signal transduction and in lipid biosynthesis in cells. This enzyme localizes to the endoplasmic reticulum. This gene is located in the class III region of the human major histocompatibility complex. Alternative splicing results in two transcript variants encoding the same protein.
Specifications
| AGPAT1 | |
| Polyclonal | |
| Unconjugated | |
| AGPAT1 | |
| 1-acylglycerol-3-phosphate O-acyltransferase 1; 1-acylglycerol-3-phosphate O-acyltransferase 1 (acetoacetly Coenzyme A thiolase); 1-acylglycerol-3-phosphate O-acyltransferase 1 (lysophosphatidic acid acyltransferase, alpha); 1-acyl-sn-glycerol-3-phosphate acyltransferase alpha; 1-AGP; 1-AGP acyltransferase 1; 1-AGPAT; 1-AGPAT 1; 1-AGPAT1; Agpat1; AW047140; G15; LPAATA; LPAAT-alpha; Lysophosphatidic acid acyltransferase alpha; lysophospholipid acyltransferase; Protein G15 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10554, 406165, 55979 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| 1.05 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O35083, Q99943 | |
| AGPAT1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 204-283 of human AGPAT1 (NP_0064021). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction