missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGPS Antibody (AGPS-03), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62219AF594
This item is not returnable.
View return policy
Description
AGPS Monoclonal specifically detects AGPS in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| AGPS | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| AGPS | |
| recombinant human AGPS (amino acids 158-384) | |
| 0.1 mL | |
| 8540 | |
| Human | |
| IgG2a |
| Western Blot, Flow Cytometry | |
| AGPS-03 | |
| Western Blot, Flow Cytometry | |
| ADAP-S, ADAS, ADHAPSAlkylglycerone-phosphate synthase, ADPS, Aging-associated gene 5 protein, aging-associated protein 5, ALDHPSY, Alkyl-DHAP synthase, alkyldihydroxyacetonephosphate synthase, peroxisomal, alkylglycerone phosphate synthase, DKFZp762O2215, EC 2.5.1.26, FLJ99755 | |
| Mouse | |
| Protein A purified | |
| Primary | |
| The antibody MEM-154 reacts with an epitope on CD16 antigen that is residing in proximity to FG loop (probably BC or C'E loop). CD16 is a low affinity receptor for aggregated IgG (FcgammaRIII antigen). The antibody MEM-154 reacts with CD16+ granulocytes. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction