missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGR2 Mouse anti-Human, Clone: UMAB245, liquid, UltraMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies UM800137
This item is not returnable.
View return policy
Description
AGR2 (anterior gradient protein 2) and AGR3 are members of the anterior gradient homolog family. They are secreted cytoplasmic proteins which are involved in metastasis induction and p53 tumour supressor inhibition. They may serve as molecular markers and potential therapeutic targets for hormone-responsive breast tumours. AGR2 is ubiquitously expressed with upregulated expression in prostate cancer, breast cancer, lung cancer, renal carcinomas and endometrial carcinomas. AGR2 expression is positively correlated with that of the estrogen receptor (ER) and is negatively correlated with that of the epidermal growth factor receptor (EGFR).Specifications
| AGR2 | |
| Monoclonal | |
| Unconjugated | |
| AGR2 | |
| AG-2, AG2, GOB-4, HAG-2, HEL-S-116, HPC8, PDIA17, XAG-2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10551 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| UMAB245 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O95994 | |
| AGR2 | |
| Human recombit protein fragment corresponding to amino acids 21-175 of human AGR2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction