missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agrin Antibody (Agr 33), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-12358C
This item is not returnable.
View return policy
Description
Agrin Monoclonal antibody specifically detects Agrin in Mouse, Rat samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Agrin | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Agr 33 | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| agrin, agrin proteoglycan | |
| Recombinant rat agrin, C-terminal construct. | |
| 0.2 mL | |
| Extracellular Matrix, Neuroscience | |
| 375790 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction