missing translation for 'onlineSavingsMsg'
Learn More

Agroclavine, TRC

Product Code. 30135307
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135307 25 mg 25mg
30134572 10 mg 10mg
17990535 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135307 Supplier Toronto Research Chemicals Supplier No. TRCA43004025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 548-42-5
Molecular Formula C16 H18 N2
Molecular Weight (g/mol) 238.33
Synonym Ergoline, 8,9-didehydro-6,8-dimethyl-, Agroclavine (6CI), 8,9-Didehydro-6,8-dimethylergoline, Indolo[4,3-fg]quinoline, ergoline deriv., (-)-Agroclavine, Agroclavin, NSC 93132
IUPAC Name (6aR,10aR)-7,9-dimethyl-6,6a,8,10a-tetrahydro-4H-indolo[4,3-fg]quinoline
SMILES CN1CC(=C[C@H]2[C@H]1Cc3c[nH]c4cccc2c34)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H18N2/c1-10-6-13-12-4-3-5-14-16(12)11(8-17-14)7-15(13)18(2)9-10/h3-6,8,13,15,17H,7,9H2,1-2H3/t13-,15-/m1/s1
Formula Weight 238.147
Percent Purity >95%
Chemical Name or Material Agroclavine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.