missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AgRP/ART Antibody (03), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23482
This item is not returnable.
View return policy
Description
AgRP/ART Monoclonal antibody specifically detects AgRP/ART in Human samples. It is validated for ELISA
Specifications
| AgRP/ART | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 3 | |
| ELISA 1:1000-1:2000 | |
| agouti related protein homolog (mouse), agouti-related protein, Agrt, AGRTMGC118963, ARTagouti (mouse) related protein, ASIP2 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human AgRP/ART (Accession#: NP_001129.1; Ser85-Thr132). | |
| 100 μg | |
| Cardiovascular Biology | |
| 181 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction