missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AgRP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118934
This item is not returnable.
View return policy
Description
AgRP Polyclonal Antibody for IHC (P)
Agouti-related protein (AgRP), also called agouti-related peptide, is a neuropeptide produced in the brain by the AgRP/NPY neuron. In humans, the agouti-related peptide is encoded by the AGRP gene. This gene encodes an antagonist of the melanocortin-3 and melanocortin-4 receptor. It appears to regulate hypothalamic control of feeding behavior via melanocortin receptor and/or intracellular calcium regulation, and thus plays a role in weight homeostasis. Mutations in this gene have been associated with late on-set obesity.
Specifications
| AgRP | |
| Polyclonal | |
| Unconjugated | |
| Agrp | |
| agouti related neuropeptide; agouti related protein; agouti related protein homolog; agouti-related protein; Agrp; AGRT; ART; ASIP2; orexigenic neuropeptide | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 181 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O00253 | |
| Agrp | |
| Recombinant protein Agouti Related Protein. The antigen corresponds to amino acid range 20-130 of the target protein. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction