missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGTPBP1 Antibody (OTI9A3), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71734C
This item is not returnable.
View return policy
Description
AGTPBP1 Monoclonal specifically detects AGTPBP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| AGTPBP1 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| AGTPBP1 | |
| Recombinant protein fragment corresponding to amino acids 368-753 of human AGTPBP1 (NP_056054) produced in E.coli. | |
| 0.1 mL | |
| 23287.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI9A3 | |
| Western Blot, Immunohistochemistry | |
| ATP/GTP binding protein 1, ATP/GTP-binding protein 1, CCP1, DKFZp686M20191, EC 3.4.17, EC 3.4.17.-, KIAA1035NNA1cytosolic carboxypeptidase 1, Nervous system nuclear protein induced by axotomy protein 1 homolog, Nna1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction