missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGTR-2 Antibody (364805), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3659W
This item is not returnable.
View return policy
Description
AGTR-2 Monoclonal antibody specifically detects AGTR-2 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| AGTR-2 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 364805 | |
| Flow Cytometry, CyTOF-ready | |
| Angiostensin Receptor, angiotensin II receptor, type 2, Angiotensin II type-2 receptor, angiotensin receptor 2, AT2ATGR2, MRX88type-2 angiotensin II receptor | |
| HEK293 human embryonic kidney cell line transfected with human AGTR-2, Met1-Ser363, Accession # P50052 | |
| 0.1 mL | |
| Apoptosis, GPCR | |
| 186 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction