missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGTRAP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals H00057085-D01P
This item is not returnable.
View return policy
Description
AGTRAP Polyclonal antibody specifically detects AGTRAP in Human samples. It is validated for Western Blot
Specifications
| AGTRAP | |
| Polyclonal | |
| Western Blot 1:500 | |
| NP_001035284.1 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| angiotensin II receptor-associated protein, AT1 receptor-associated protein, ATI receptor-associated protein, MGC29646, type I receptor-associated protein, type-1 angiotensin II receptor-associated protein | |
| AGTRAP (NP_001035284.1, 1 a.a. - 152 a.a.) full-length human protein. MELPAVNLKVILLGHWLLTTWGCIVFSGSYAWANFTILALGVWAVAQRDSIDAISMFLGGLLATIFLDIVHISIFYPRVSLTDTGRFGVGMAILSLLLKPLSCCFVYHMYRERGGFLGSSQDRSAYQTIDSAEAPADPFAVPEGRSQDARGY | |
| 0.1 mg | |
| Cardiovascular Biology, GPCR, Signal Transduction | |
| 57085 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction