missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ AGXT Antibody (2D10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-31960-100ul
This item is not returnable.
View return policy
Description
AGXT Monoclonal antibody specifically detects AGXT in Human samples. It is validated for Flow Cytometry,ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| AGXT | |
| Monoclonal | |
| 1 mg/mL | |
| PBS (pH 7.4), 10% Glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 2D10 | |
| Unconjugated | |
| AGT1L-alanine: glyoxylate aminotransferase 1, AGThepatic peroxisomal alanine:glyoxylate aminotransferase, AGXT1, alanine-glyoxylate aminotransferase, Alanine--glyoxylate aminotransferase, EC 2.6.1.44, EC 2.6.1.51, PH1, serine:pyruvate aminotransferase, SPATserine-pyruvate aminotransferase, SPTserine--pyruvate aminotransferase, TLH6 | |
| Recombinant AGXT (330-392aa) purified from E. coli | |
| 100 μL | |
| Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction | |
| 189 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction