missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGXT Antibody (S02-7A4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19562-100ul
This item is not returnable.
View return policy
Description
AGXT Monoclonal antibody specifically detects AGXT in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, RNA Inhibition, Sandwich ELISA
Specifications
| AGXT | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Inhibition Assays, Sandwich ELISA | |
| S02-7A4 | |
| Unconjugated | |
| AGT1L-alanine: glyoxylate aminotransferase 1, AGThepatic peroxisomal alanine:glyoxylate aminotransferase, AGXT1, alanine-glyoxylate aminotransferase, Alanine--glyoxylate aminotransferase, EC 2.6.1.44, EC 2.6.1.51, PH1, serine:pyruvate aminotransferase, SPATserine-pyruvate aminotransferase, SPTserine--pyruvate aminotransferase, TLH6 | |
| Recombinant protein of human AGXT (Uniprot # P21549) | |
| 100 μg | |
| Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction | |
| 189 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction