Learn More
AGXT Mouse anti-Human, Unconjugated, Clone: 3E12H1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67402-1-IG-20UL
This item is not returnable.
View return policy
Description
The AGXT antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AGXT. This antibody recognizes human antigen. The AGXT antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| AGXT | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, ELISA | |
| 3E12H1 | |
| Unconjugated | |
| 20 μL | |
| 189 | |
| IgG1 |