Learn More
Invitrogen™ AGXT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597036
Description
Immunogen sequence: DSVASLGGTP LYMDRQGIDI LYSGSQKALN APPGTSLISF SDKAKKKMYS RKTKPFSFYL DIKWLANFWG CDDQPRMYHH TIPVISLYSL RESLALIAEQ GLENSWRQHR EAAAYLHGRL QALGLQLFVK DPALRLPTVT TVAVPAGYDW RDIVSYVIDH FDIEIMGGLG PSTGKVLRIG LLGCNATREN VDRVTEALRA ALQHCPKKKL; Positive Samples: LO2, 293T; Cellular Location: Mitochondrion, Peroxisome.
This gene is expressed only in the liver and the encoded protein is localized mostly in the peroxisomes, where it is involved in glyoxylate detoxification. Mutations in this gene, some of which alter subcellular targetting, have been associated with type I primary hyperoxaluria.
Specifications
| AGXT | |
| Polyclonal | |
| Unconjugated | |
| AGXT | |
| AGT; Agt1; Agxt; Agxt1; alanine-glyoxylate aminotransferase; Alanine--glyoxylate aminotransferase; alanine-glyoxylate aminotransferase 1; angiotensin receptor 2; hepatic peroxisomal alanine:glyoxylate aminotransferase; L-alanine: glyoxylate aminotransferase 1; PH1; serine:pyruvate aminotransferase; serine:pyruvate aminotransferase SPT; serine:pyruvate/alanine:glyoxylate aminotransferase; serine-pyruvate aminotransferase; Serine--pyruvate aminotransferase; serine--pyruvate aminotransferase, mitochondrial; serine--pyruvate aminotransferase, mitochondrial; serine--pyruvate aminotransferase, peroxisomal; serine--pyruvate aminotransferase, peroxisomal; serine--pyruvate aminotransferase; serine--pyruvate aminotransferase, mitochondrial; serine--pyruvate aminotransferase, peroxisomal; Spat; SPT; TLH6 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 189 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.82 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P21549 | |
| AGXT | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 183-392 of human AGXT (NP_0000211). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.