missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AGXT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598871
This item is not returnable.
View return policy
Description
AGXT2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
The protein encoded by this gene is a class III pyridoxal-phosphate-dependent mitochondrial aminotransferase. It catalyzes the conversion of glyoxylate to glycine using L-alanine as the amino donor.
Specifications
| AGXT2 | |
| Polyclonal | |
| Unconjugated | |
| Agxt2 | |
| (R)-3-amino-2-methylpropionate--pyruvate transaminase; AGT 2; Agt2; Agxt2; AI303810; AI663818; alanine-glyoxylate aminotransferase 2; alanine--glyoxylate aminotransferase 2; alanine--glyoxylate aminotransferase 2, mitochondrial; beta-ALAAT II; Beta-alanine-pyruvate aminotransferase; DAIBAT; D-AIBAT | |
| Rabbit | |
| Protein G | |
| RUO | |
| 64902 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9BYV1 | |
| Agxt2 | |
| Recombinant Human Alanine--glyoxylate aminotransferase 2, mitochondrial protein (196-328AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction