missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AHI1 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78206AF700
This item is not returnable.
View return policy
Description
AHI1 Polyclonal antibody specifically detects AHI1 in Human samples. It is validated for Western Blot
Specifications
| AHI1 | |
| Polyclonal | |
| Western Blot | |
| Abelson helper integration site, Abelson helper integration site 1, Abelson helper integration site 1 protein homolog, AHI-1, contatins SH3 and WD40 domains, dJ71N10.1, DKFZp686J1653, FLJ14023, FLJ20069, JBTS3, jouberin, ORF1 | |
| The immunogen for this product maps to a region between residue 1146 and 1196 of human Abelson Helper Integration site 1 using the numbering given in entry NP_001128302.1 (GeneID 54806). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 54806 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction