Learn More
AHI1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Product Code. 16891585 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16891585 150 μL 150µL
16801595 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16891585 Supplier Proteintech Supplier No. 220451AP150UL
Rabbit Polyclonal Antibody
This gene is apparently required for both cerebellar and cortical development in humans. This gene mutations cause specific forms of Joubert syndrome-related disorders. Joubert syndrome (JS) is a recessively inherited developmental brain disorder with several identified causative chromosomal loci. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
Antigen AHI1
Applications Western Blot
Classification Polyclonal
Concentration 0.18 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene AHI1
Gene Accession No. Q8K3E5, Q8N157
Gene Alias AHI 1, AHI1, JBTS3, Jouberin, ORF1
Gene Symbols Ahi1
Host Species Rabbit
Immunogen AHI1 Fusion Protein Ag16885
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 52906, 54806
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG