missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AHNAK Antibody (EM-09), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62204UV
This item is not returnable.
View return policy
Description
AHNAK Monoclonal specifically detects AHNAK in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
| AHNAK | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| AHNAK | |
| Bacterially expressed fragment of N-terminal domain of human AHNAK1. | |
| 0.1 mL | |
| Primary | |
| The antibody C-51 reacts with Cytokeratin 8 (52.5 kDa). Cytokeratins are a member of intermediate filaments subfamily represented in epithelial tissues. | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| EM-09 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
| AHNAK nucleoprotein, AHNAK nucleoprotein (desmoyokin), AHNAK-related, AHNAKRS, Desmoyokin, MGC5395, neuroblast differentiation-associated protein AHNAK, PM227 | |
| Mouse | |
| Protein A purified | |
| Neuroscience | |
| 79026 | |
| Human, Mouse | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction