Learn More
AHNAK Antibody (EM-09), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62204H
This item is not returnable.
View return policy
Description
AHNAK Monoclonal specifically detects AHNAK in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| AHNAK | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| AHNAK | |
| Bacterially expressed fragment of N-terminal domain of human AHNAK1. | |
| 0.1 mL | |
| Primary | |
| The antibody C-51 reacts with Cytokeratin 8 (52.5 kDa). Cytokeratins are a member of intermediate filaments subfamily represented in epithelial tissues. | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| EM-09 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen | |
| AHNAK nucleoprotein, AHNAK nucleoprotein (desmoyokin), AHNAK-related, AHNAKRS, Desmoyokin, MGC5395, neuroblast differentiation-associated protein AHNAK, PM227 | |
| Mouse | |
| Protein A purified | |
| Neuroscience | |
| 79026 | |
| Human, Mouse | |
| IgG1 |