Learn More
AHNAK Antibody (EM-09), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-62204H
Product Code. 18224804
  • € 522.00 / 0.10mL
Description
AHNAK Monoclonal specifically detects AHNAK in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AHNAK
Monoclonal
HRP
PBS with No Preservative
AHNAK
Bacterially expressed fragment of N-terminal domain of human AHNAK1.
0.1 mL
Primary
The antibody C-51 reacts with Cytokeratin 8 (52.5 kDa). Cytokeratins are a member of intermediate filaments subfamily represented in epithelial tissues.
Store at 4°C in the dark.
Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)
EM-09
Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen
AHNAK nucleoprotein, AHNAK nucleoprotein (desmoyokin), AHNAK-related, AHNAKRS, Desmoyokin, MGC5395, neuroblast differentiation-associated protein AHNAK, PM227
Mouse
Protein A purified
Neuroscience
79026
Human, Mouse