missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AHNAK Antibody (PSH01-11), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32010
This item is not returnable.
View return policy
Description
AHNAK Monoclonal antibody specifically detects AHNAK in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| AHNAK | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100-1:250, Immunohistochemistry-Paraffin 1:1000 | |
| AHNAK nucleoprotein, AHNAK nucleoprotein (desmoyokin), AHNAK-related, AHNAKRS, Desmoyokin, MGC5395, neuroblast differentiation-associated protein AHNAK, PM227 | |
| Recombinant protein within human AHNAK aa 451-900/ 5,890. (Uniprot: Q09666) | |
| 100 μL | |
| Neuroscience | |
| 79026 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| PSH01-11 | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction