Learn More
Invitrogen™ AHNAK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551475
Description
Immunogen sequence: TKYSQESWFK MPKFRMPSLR RSFRDRGGAG KLEVAQTQAP AATGGEAAAK VKEFLVSGSN VEAAMSLQLP EADAEVTASE SKSSTDILRC DLDSTGLKLH LSTAGMTGDE LSTSEVRIHP SKGPLPFQMP GMRLPETQVL PGEIDETP Highest antigen sequence identity to the following orthologs: Mouse - 36%, Rat - 31%.
The specific function of AHNAK2 is not yet known.
Specifications
| AHNAK2 | |
| Polyclonal | |
| Unconjugated | |
| AHNAK2 | |
| AHNAK nucleoprotein 2; AHNAK2; AI450948; C14orf78; Gm1185; Gm72; KIAA2019; Protein AHNAK2; RGD1309696 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 113146 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IVF2 | |
| AHNAK2 | |
| Recombinant protein corresponding to Human AHNAK2. Recombinant protein control fragment (Product #RP-88995). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.