missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AHSA1 Antibody (25F2.D10), Alexa Fluor™ 350, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12884AF350
This item is not returnable.
View return policy
Description
AHSA1 Monoclonal specifically detects AHSA1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| AHSA1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| AHSA1 | |
| Mouse Aha1 | |
| Primary | |
| Detects approx 38kDa. Can run up to 45kDa on SDS Page. | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 25F2.D10 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| activator of 90 kDa heat shock protein ATPase homolog 1, AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast), C14orf3, chromosome 14 open reading frame 3, p38AHA1 | |
| Rat | |
| 0.1 mL | |
| 10598 | |
| Human, Mouse, Rat | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction