missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AHSA1 Antibody (4H9-D7), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12891G
This item is not returnable.
View return policy
Description
AHSA1 Monoclonal antibody specifically detects AHSA1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| AHSA1 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| 4H9-D7 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| activator of 90 kDa heat shock protein ATPase homolog 1, AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast), C14orf3, chromosome 14 open reading frame 3, p38AHA1 | |
| Human Aha1, his tagged | |
| 0.1 mL | |
| Signal Transduction | |
| 10598 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction