missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ AHSA1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29181
This item is not returnable.
View return policy
Description
AHSA1 Polyclonal antibody specifically detects AHSA1 in Human samples. It is validated for Immunoprecipitation
Specifications
| AHSA1 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| Unconjugated | |
| activator of 90 kDa heat shock protein ATPase homolog 1, AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast), C14orf3, chromosome 14 open reading frame 3, p38AHA1 | |
| Between 288 and 338 | |
| 100 μg | |
| Signal Transduction | |
| 10598 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction