missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AHSA1 Antibody (OTI1D2), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71608B
This item is not returnable.
View return policy
Description
AHSA1 Monoclonal specifically detects AHSA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated.
Specifications
| AHSA1 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| AHSA1 | |
| Recombinant protein fragment corresponding to amino acids 1-203 of human AHSA1 (NP_036243) produced in E.coli. | |
| 0.1 mL | |
| 10598.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunoassay | |
| OTI1D2 | |
| Western Blot, Immunohistochemistry, Knockout Validated | |
| activator of 90 kDa heat shock protein ATPase homolog 1, AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast), C14orf3, chromosome 14 open reading frame 3, p38AHA1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction