missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AHSA2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17180874 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17180874 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17180874 Supplier Invitrogen™ Supplier No. PA5113017

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AHSA2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Co-chaperone that stimulates HSP90 ATPase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AHSA2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Ahsa2
Gene Accession No. Q719I0, Q8N9S3
Gene Alias Activator of 90 kDa heat shock protein ATPase homolog 2; Activator of heat shock protein ATPase homolog 2; AHA1, Activator of heat shock 90kDa protein ATPase homolog 2 (yeast); AHA1, Activator of heat shock protein ATPase 2; AHA1, Activator of heat shock protein ATPase homolog 2; Ahsa2; Hch1
Gene Symbols Ahsa2
Host Species Rabbit
Immunogen Recombinant Human Putative activator of 90 kDa heat shock protein ATPase homolog 2 protein (1-137AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130872, 268390, 305577
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.