Learn More
AIF-1/Iba1 Antibody (031), mFluor Violet 500 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-90271MFV500
Product Code. 18477789
  • € 674.00 / 0.10mL
Description
AIF-1/Iba1 Monoclonal antibody specifically detects AIF-1/Iba1 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)