missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIF-1/Iba1 Antibody (rAIF1/1909), Novus Biologicals™
Mouse Monoclonal Antibody
€ 274.05 - € 480.90
Specifications
| Antigen | AIF-1/Iba1 |
|---|---|
| Clone | rAIF1/1909 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18661638
|
Novus Biologicals
NBP2-75755-100ug |
100 μg |
€ 509.00 € 480.90 / 100µg Save € 28.10 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18637288
|
Novus Biologicals
NBP2-75755-20ug |
20 μg |
€ 290.00 € 274.05 / 20µg Save € 15.95 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AIF-1/Iba1 Monoclonal antibody specifically detects AIF-1/Iba1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Protein ArraySpecifications
| AIF-1/Iba1 | |
| 0.2 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| Unconjugated | |
| Mouse | |
| Autoimmune Diseases, Cancer, Cell Biology, Cell Cycle and Replication, Immunology, Inflammation, Lipid and Metabolism, Microglia Markers, Neuroscience | |
| 10 mM PBS with 0.05% BSA | |
| AIF-1, allograft inflammatory factor 1, G1, IBA1, IbaI, interferon gamma responsive transcript, Ionized calcium-binding adapter molecule 1, IRT1, IRT-1, Protein G1 | |
| Purified recombinant human AIF-1/Iba1 protein (around aa 1-146) (exact sequence is proprietary) (Uniprot: P55008) | |
| Primary | |
| Store at 4°C. |
| rAIF1/1909 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P55008 | |
| 199 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title