missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIF Antibody (3C11), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50606AF350
This item is not returnable.
View return policy
Description
AIF Monoclonal specifically detects AIF in Human samples. It is validated for Western Blot.
Specifications
| AIF | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| AIFM1 | |
| AIF-His fusion protein | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| 3C11 | |
| Western Blot | |
| AIFapoptosis-inducing factor 1, mitochondrial, apoptosis-inducing factor, mitochondrion-associated, 1, COXPD6, MGC111425, PDCD8striatal apoptosis-inducing factor, programmed cell death 8 (apoptosis-inducing factor), Programmed cell death protein 8 | |
| Mouse | |
| Protein A purified | |
| Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, Inhibitors Activators and Regulators, Mitochondrial Markers, Tumor Suppressors | |
| 9131.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction