missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIF1L Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97372AF350
This item is not returnable.
View return policy
Description
AIF1L Polyclonal antibody specifically detects AIF1L in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| AIF1L | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 83543 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 350 | |
| allograft inflammatory factor 1-like, C9orf58, chromosome 9 open reading frame 58, IBA2FLJ12783, ionized calcium binding adapter molecule 2, Ionized calcium-binding adapter molecule 2, MGC29466 | |
| Produced in rabbits immunized with E. coli-derived Human AIF1L fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction