missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIM2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15313
This item is not returnable.
View return policy
Description
AIM2 Polyclonal specifically detects AIM2 in Human samples. It is validated for Western Blot.
Specifications
| AIM2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| absent in melanoma 2PYHIN4, interferon-inducible protein AIM2 | |
| Rabbit | |
| 39 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.75mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry-Paraffin | |
| O14862 | |
| AIM2 | |
| Recombinant protein encompassing a sequence within the center region of human AIM2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 9447 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction