missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIM2 Mouse anti-Human, Unconjugated, Clone: 3E12A8, Proteintech
Mouse Monoclonal Antibody
€ 149.35 - € 422.30
Specifications
| Antigen | AIM2 |
|---|---|
| Clone | 3E12A8 |
| Concentration | 1000 μg/mL |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16829185
|
Proteintech
66902-1-IG-20UL |
20 μL |
€ 149.35
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16819185
|
Proteintech
66902-1-IG-150UL |
150 μL |
€ 422.30
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The AIM2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AIM2. This antibody recognizes human, mouse, rat, rabbit antigen. The AIM2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| AIM2 | |
| 1000 μg/mL | |
| Unconjugated | |
| Human, Mouse, Rat, Rabbit | |
| IgG1 |
| 3E12A8 | |
| Monoclonal | |
| Mouse | |
| 9447 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title