missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AIM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112354
This item is not returnable.
View return policy
Description
AIM2 Polyclonal Antibody for Western Blot, IHC (P), ICC/IF, ELISA
AIM2 is a member of the IFI20X /IFI16 family. It plays a putative role in tumorigenic reversion and may control cell proliferation. Interferon-gamma induces expression of AIM2.
Specifications
| AIM2 | |
| Polyclonal | |
| Unconjugated | |
| AIM2 | |
| Absent in melanoma 2; AIM2; Gm1313; Ifi210; ifi-210; Interferon-inducible protein 210; interferon-inducible protein AIM2; Interferon-inducible protein p210; PYHIN4; RP11-520H16.3 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 9447 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O14862 | |
| AIM2 | |
| Recombinant Human Interferon-inducible protein AIM2 protein (81-326AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction