missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AIMP2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17949052
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17949052 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17949052 Supplier Invitrogen™ Supplier No. PA5144670

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive control: Hela cell lysate, Jurkat cell lysate, A549 cell lysate, SH-SY5Y cell lysate, human kidney tissue lysate, rat kidney tissue lysate, mouse kidney tissue lysate, human testis tissue, mouse kidney tissue, SH-SY5Y. Predicted band size: 35 kDa Subcellular Location: Cytoplasm, cytosol, Nucleus.

JTV1 is probable core protein of the multisynthetase complex that serves as a template for the assembly of the supramolecular structure. This protein Mediates ubiquitination of FUBP1 and its degradation by the proteasome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AIMP2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene AIMP2
Gene Accession No. Q13155, Q32PX2, Q8R010
Gene Alias AA407136; AA407225; AIMP2; Aimp2(p38); Aminoacyl tRNA synthase complex-interacting multifunctional protein 2; aminoacyl tRNA synthetase complex interacting multifunctional protein 2; aminoacyl tRNA synthetase complex-interacting multifunctional protein 2; ARS interacting multifunctional protein 2; ARS-interacting multi-functional protein 2; JTV1; JTV-1; multisynthase complex auxiliary component p38; multisynthetase complex auxiliary component p38; P38; PRO0992; Protein JTV-1
Gene Symbols AIMP2
Host Species Rabbit
Immunogen Recombinant protein within human AIMP2 aa 71-320/320.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231872, 288480, 7965
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.