missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aiolos/IKZF3 Antibody (914821) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8625JF585
This item is not returnable.
View return policy
Description
Aiolos/IKZF3 Monoclonal antibody specifically detects Aiolos/IKZF3 in Human samples. It is validated for Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| Aiolos/IKZF3 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| AIO, AIOLOS, IKAROS family zinc finger 3 (Aiolos), Ikaros family zinc finger protein 3, zinc finger protein Aiolos, ZNFN1A3subfamily 1A, 3 (Aiolos) | |
| E. coli-derived recombinant human Aiolos/IKZF3, Met1-Gly54, Accession # Q9UKT9 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 914821 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Chromatin Research | |
| 22806 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction