missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aiolos/IKZF3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16938
This item is not returnable.
View return policy
Description
Aiolos/IKZF3 Polyclonal specifically detects Aiolos/IKZF3 in Human samples. It is validated for Western Blot.
Specifications
| Aiolos/IKZF3 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris-Glycine (pH7), 20% Glycerol with 0.01% Thimerosal | |
| AIO, AIOLOS, IKAROS family zinc finger 3 (Aiolos), Ikaros family zinc finger protein 3, zinc finger protein Aiolos, ZNFN1A3subfamily 1A, 3 (Aiolos) | |
| Rabbit | |
| 58 kDa | |
| 0.1 mL | |
| Chromatin Research | |
| 22806 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.74mg/mL | |
| Western Blot 1:500-1:3000, Simple Western reported by internal validation | |
| Q9UKT9 | |
| IKZF3 | |
| Recombinant protein encompassing a sequence within the center region of human IKZF3. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction