missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AIP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587148
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
This gene encodes a member of the NEDD4-like protein family. The family of proteins is known to possess ubiquitin-protein ligase activity. The encoded protein contains 4 tandem WW domains. The WW domain is a protein motif consisting of 35 to 40 amino acids and is characterized by 4 conserved aromatic residues. The WW domain may mediate specific protein-protein interactions. Three alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
| AIP2 | |
| Polyclonal | |
| Unconjugated | |
| WWP2 | |
| 1300010O06Rik; AA690238; AIP2; atrophin-1 interacting protein 2; Atrophin-1-interacting protein 2; AW554328; HECT-type E3 ubiquitin transferase WWP2; id:ibd1121; NEDD4-like E3 ubiquitin-protein ligase WWP2; wu:fo87e10; WW domain containing E3 ubiquitin protein ligase 2; WW domain-containing protein 2; WW domain-containing protein 4; WWP2; WWp2-like; zgc:154036 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11060 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O00308 | |
| WWP2 | |
| Recombinant fusion protein of human WWP2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction