missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIPL1 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76953V
This item is not returnable.
View return policy
Description
AIPL1 Polyclonal antibody specifically detects AIPL1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| AIPL1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 405 | |
| AIPL2, aryl hydrocarbon receptor interacting protein-like 1, aryl hydrocarbon receptor-interacting protein-like 1, aryl-hydrocarbon-interacting protein-like 1, LCA4 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of the human Aipl1. The immunogen is located within amino acids 290 - 340 of Aipl1. Amino Acid Squence: RLLENRMAEKQEEERLR | |
| 0.1 mL | |
| Vision | |
| 23746 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction