Learn More
Abeba™ Air-Cushion 32300 Shoes
Product Code. 15659637
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Product Code. 15659637
Brand: Abeba™ 32300/35
Only null left
Add to basket
Add to basket
Air cushion PU-sole with high slip resistance. Sanitized™ treated.
  • CE EN ISO 20347:2012 O1 SRA
  • Smooth leather, perforated
  • White
  • Slip resistance PU outsole sole
  • Replaceable insole (article no. 3551)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Lining microfiber, respiration absorbing
  • Instep area with elastic band
Specifications
Certifications/Compliance CE EN ISO 20347:2012 O1 SRA
Color White
Liner Material Microfiber
Material Leather
Size 35
Sole Features Slip-resistant
Sole Material Polyurethane
Style 32300