Learn More
Aalborg™ Air Mass Flowmeter
Brand:  Aalborg™ GFM17A-VAL6-A0/3AB-07-SS
Additional Details : Weight : 0.70000kg
 View more versions of this product
Product Code. 11872170
  • € 980.50 / Each
Estimated Shipment: 05-08-2024
to see stock.
Specifications
Mass Flowmeter
0 to 1 LPM
12 V to 200 mA max
Full scale
±5%
NIST calibration certificate
±1.5 %
5°C to 50°C