missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIRE Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22517-1-AP-150UL
This item is not returnable.
View return policy
Description
The autoimmune regulator (AIRE) is a protein that in humans is encoded by the AIRE gene. This gene encodes a transcriptional regulator that forms nuclear bodies and interacts with the transcriptional coactivator CREB binding protein. The encoded protein plays an important role in immunity by regulating the expression of autoantigens and negative selection of autoreactive T-cells in the thymus. Mutations in this gene cause the rare autosomal-recessive systemic autoimmune disease termed autoimmune polyendocrinopathy with candidiasis and ectodermal dystrophy (APECED).Specifications
| AIRE | |
| Polyclonal | |
| Unconjugated | |
| AIRE | |
| AIRE, AIRE1, APECED, APECED protein, APS1, APSI, autoimmune regulator, PGA1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11634, 326 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43918, Q9Z0E3 | |
| AIRE | |
| AIRE Fusion Protein Ag18249 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction