Learn More
Uvex™ Airwing B Safety Helmet
A lightweight helmet with an attractive design.
POA
Specifications
Certifications/Compliance EN 397
Material HDPE
Model 9762
Product Type Industrial Safety Helmet
Size (Metric) 51 to 61 cm
View More Specs
Description
Description
  • Lightweight safety helmet made of polyethylene
  • Three variable air vents, one ventilation zone on the forehead and two at the back, for perfect air circulation
  • 6-point textile suspension harness
  • Side slots for attaching ear defenders or visors
  • Conventional suspension harness
Specifications
Specifications
EN 397
9762
51 to 61 cm
HDPE
Industrial Safety Helmet
Ventilated