Learn More
Uvex™ Airwing B Safety Helmet
A lightweight helmet with an attractive design.
POA
Specifications
Certifications/Compliance | EN 397 |
---|---|
Material | HDPE |
Model | 9762 |
Product Type | Industrial Safety Helmet |
Size (Metric) | 51 to 61 cm |
Description
- Lightweight safety helmet made of polyethylene
- Three variable air vents, one ventilation zone on the forehead and two at the back, for perfect air circulation
- 6-point textile suspension harness
- Side slots for attaching ear defenders or visors
- Conventional suspension harness
Specifications
EN 397 | |
9762 | |
51 to 61 cm |
HDPE | |
Industrial Safety Helmet | |
Ventilated |