missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AK1BA Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

€ 212.00 - € 514.00

Specifications

Antigen AK1BA
Concentration 0.5 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17790344
GREENER_CHOICE
View Documents
Invitrogen™
PA5118936
200 μL
€ 514.00
200µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17733654
View Documents
Invitrogen™
PA5122255
20 μL
€ 212.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AK1BA Polyclonal Antibody for Western Blot, ICC/IF, IHC

Catalyzes the NADPH-dependent reduction of a wide variety of carbonyl-containing compounds to their corresponding alcohols. Displays strong enzymatic activity toward all-trans-retinal, 9-cis-retinal, and 13-cis-retinal. Plays a critical role in detoxifying dietary and lipid-derived unsaturated carbonyls, such as crotonaldehyde, 4-hydroxynonenal, trans-2-hexenal, trans-2,4-hexadienal and their glutathione-conjugates carbonyls (GS-carbonyls). Displays no reductase activity towards glucose.
Specifications

Specifications

AK1BA
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Human, Rat
AKR1B10
AKR1B10; AKR1B11; Aldo-keto reductase family 1 member B10
AKR1B10
Primary
Antigen affinity chromatography, Protein A
0.5 mg/mL
Immunohistochemistry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
O60218
296972, 57016
Recombinant protein Aldose Reductase Like Protein 1. The antigen corresponds to amino acid range 1-316 of the target protein.
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.