Learn More
AK2 Mouse anti-Human, CoraLite 594, Clone: 3H2C10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-66127-100UL
Description
The AK2 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human AK2. This antibody recognizes human antigen. The AK2 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™594.
Specifications
| AK2 | |
| Monoclonal | |
| 2000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| 3H2C10 | |
| CoraLite 594 | |
| 100 μL | |
| 204 | |
| IgG2a |