Learn More
AK2 Mouse anti-Human, CoraLite 594, Clone: 3H2C10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL594-66127-100UL
Product Code. 16810976
  • € 450.00 / 100µL
Description
The AK2 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human AK2. This antibody recognizes human antigen. The AK2 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™594.
Specifications
Specifications
AK2
Monoclonal
2000 μg/mL
Mouse
Primary
Human
Immunofluorescence, Flow Cytometry (Intracellular Staining)
3H2C10
CoraLite 594
100 μL
204