missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AK3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38109-20ul
This item is not returnable.
View return policy
Description
AK3 Polyclonal antibody specifically detects AK3 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| AK3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| Adenylate kinase 3 alpha-like 1, adenylate kinase 3 like 1, adenylate kinase 3adenylate kinase 6, AK 3, AK6, AKL3L, AKL3L1, EC 2.7.4, EC 2.7.4.10, FIX, GTP:AMP phosphotransferase, mitochondrial | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-227 of human AK3 (NP_057366.2).,, Sequence:, MGASARLLRAVIMGAPGSGKGTVSSRITTHFELKHLSSGDLLRDNMLRGTEIGVLAKAFIDQGKLIPDDVMTRLALHELKNLTQYSWLLDGFPRTLPQAEALDRAYQIDTVINLNVPFEVIKQRLTARWIHPASGRVYNIEFNPPKTVGIDDLTGEPLIQREDDKPETVIKRLKAYEDQTKPVLEYYQKKGVLETFSGTETNKIWPYVYAFLQTKVPQRSQKASVTP | |
| 20 μL | |
| Signal Transduction | |
| 50808 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction