missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AK3 Antibody (OTI6G4), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71874G
This item is not returnable.
View return policy
Description
AK3 Monoclonal specifically detects AK3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| AK3 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| AK3 | |
| Full length recombinant protein of human AK3 (NP_057336) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI6G4 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| Adenylate kinase 3 alpha-like 1, adenylate kinase 3 like 1, adenylate kinase 3adenylate kinase 6, AK 3, AK6, AKL3L, AKL3L1, EC 2.7.4, EC 2.7.4.10, FIX, GTP:AMP phosphotransferase, mitochondrial | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 50808.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction